missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Chelerythrine chloride
GREENER_CHOICE

Product Code. 18043912
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18043912 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18043912 Supplier Tocris Bioscience™ Supplier No. 1330/5
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cell-permeable protein kinase C inhibitor

Cell-permeable inhibitor of protein kinase C (IC50 = 660 nM); competitive with respect to the phosphate acceptor and non-competitive with respect to ATP. Has a wide range of biological activities, including antiplatelet, anti-inflammatory, antibacterial and antitumor effects. Activates MAPK pathways, independent of PKC inhibition. Inhibits binding of BclXL to Bak (IC50 = 1.5 μM) or Bad proteins and stimulates apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 3895-92-9
Quantity 5 mg
Molecular Formula C21H18ClNO4
Formula Weight 383.83
Purity >98%
Molecular Weight (g/mol) 383.83
Chemical Name or Material Chelerythrine chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.