missing translation for 'onlineSavingsMsg'
Learn More

Chelerythrine chloride, MedChemExpress

Product Code. 16497687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497687 5 mg 5mg
16475726 100 mg 100mg
16471197 50 mg 50mg
16404958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497687 Supplier Medchem Express Supplier No. HY12048
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Chelerythrine chloride is a potent, cell-permeable inhibitor of protein kinase C, with an IC50 of 660 nM. Chelerythrine chloride inhibits the Bcl-XL-Bak BH3 peptide binding with IC50 of 1.5 μM and displaces Bax from Bcl-XL. Chelerythrine chloride induces apoptosis and autophagy.

Chelerythrine chloride is a potent, cell-permeable inhibitor of protein kinase C, with an IC50 of 660 nM. Chelerythrine chloride inhibits the Bcl-XL-Bak BH3 peptide binding with IC50 of 1.5 μM and displaces Bax from Bcl-XL. Chelerythrine chloride induces apoptosis and autophagy.

Specifications

Chemical Name or Material Chelerythrine chloride
CAS 3895-92-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H18ClNO4
Quantity 5 mg
Solubility Information DMSO : 4.35 mg/mL (11.33 mM; Need ultrasonic)
SMILES C[N+]1=CC2=C(C(OC)=CC=C2C3=C1C4=CC5=C(OCO5)C=C4C=C3)OC.[Cl-]
Molecular Weight (g/mol) 383.82
Formula Weight 383.82
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.