missing translation for 'onlineSavingsMsg'
Learn More

Chelerythrine chloride, 99+%

Product Code. 15495199
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495199 25 mg 25mg
15475199 5 mg 5mg
15485199 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495199 Supplier Thermo Scientific Alfa Aesar Supplier No. J62906.MB
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3895-92-9 | C21H18ClNO4 | 383.83 g/mol

A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A natural benzophenanthridine alkaloid
  • Inhibits protein kinase C
  • Induces apoptosis in leukemia cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3895-92-9
Molecular Formula C21H18ClNO4
Molecular Weight (g/mol) 383.83
MDL Number MFCD00060717
InChI Key WEEFNMFMNMASJY-UHFFFAOYSA-M
Synonym chelerythrine chloride, chelerythrine hydrochloride, chelerythrinechloride, unii-7ic98tz0pz, 1,2-dimethoxy-12-methyl-1,3 dioxolo 4',5':4,5 benzo 1,2-c phenanthridin-12-ium chloride, chelerythrine, chloride, 7ic98tz0pz, 1,2-dimethoxy-n-methyl-1,3-benzodioxolo 5,6c phenanthridinium, 1,2-dimethoxy-12-methyl 1,3 benzodioxolo 5,6-c phenanthridinium chloride
PubChem CID 72311
SMILES [Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC

Specifications

Color Yellow to Orange
Quantity 25 mg
Merck Index 14,2051
Solubility Information Soluble in DMSO at 2mg/ml
IUPAC Name 17,18-dimethoxy-21-methyl-5,7-dioxa-21-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁴,¹⁹]henicosa-1(13),2,4(8),9,11,14,16,18,20-nonaen-21-ium chloride
Formula Weight 383.82
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Chelerythrine chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.