missing translation for 'onlineSavingsMsg'
Learn More

Chelerythrine chloride, 99+%

Product Code. 15485199
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 15485199

Brand: Thermo Scientific Alfa Aesar J62906.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3895-92-9 | C21H18ClNO4 | 383.83 g/mol

A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A natural benzophenanthridine alkaloid
  • Inhibits protein kinase C
  • Induces apoptosis in leukemia cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3895-92-9
Molecular Formula C21H18ClNO4
Molecular Weight (g/mol) 383.83
MDL Number MFCD00060717
InChI Key WEEFNMFMNMASJY-UHFFFAOYSA-M
Synonym chelerythrine chloride, chelerythrine hydrochloride, chelerythrinechloride, unii-7ic98tz0pz, 1,2-dimethoxy-12-methyl-1,3 dioxolo 4',5':4,5 benzo 1,2-c phenanthridin-12-ium chloride, chelerythrine, chloride, 7ic98tz0pz, 1,2-dimethoxy-n-methyl-1,3-benzodioxolo 5,6c phenanthridinium, 1,2-dimethoxy-12-methyl 1,3 benzodioxolo 5,6-c phenanthridinium chloride
PubChem CID 72311
SMILES [Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC

Specifications

Color Yellow to Orange
Quantity 10 mg
Merck Index 14,2051
Solubility Information Soluble in DMSO at 2mg/ml
IUPAC Name 1,2-dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridin-12-ium;chloride
Formula Weight 383.82
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material Chelerythrine chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.