Learn More
Chelerythrine chloride, 99+%
Description
A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A natural benzophenanthridine alkaloid
- Inhibits protein kinase C
- Induces apoptosis in leukemia cells
Specifications
Specifications
| Color | Yellow to Orange |
| Quantity | 5 mg |
| Merck Index | 14,2051 |
| Solubility Information | Soluble in DMSO at 2mg/ml |
| IUPAC Name | 1,2-dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridin-12-ium;chloride |
| Formula Weight | 383.82 |
| Percent Purity | ≥99% |
| Physical Form | Solid |
| Chemical Name or Material | Chelerythrine chloride |
Safety and Handling
- Chelerythrine chloride
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contain: 1,2-Dimethoxy-12-methyl[1,3]benzodioxolo[5,6-c]phenanthridinium chloride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.