Learn More
Aqualytic™ Checkit Ozone DPD Test Kit
Click to view available options
Range:
0 to 1.0mg/L
Unit Size:
Each
Description
Aqualytic Checkit Ozone DPD Test Kit, Range: 0 to 1.0mg/L, Accuracy: +/-5%, Quantity: 1Pack
Specifications
Specifications
| Accuracy | ±5% |
| Quantity | 1 Pack |
| Range | 0 to 1.0mg/L |
| Type | Checkit Ozone DPD Test Kit |