Learn More
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
Brand: Lovibond™ 147235
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
- Easy operation and exact reagent dosing
- Tablet reagents with a minimm guaranteed shelf life of 5/10 years
- Rotate continuous gradient color wheel to obtain a color match with the reacted sample
- Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
pH Value Copper |
|
6.5 to 8.4 pH 0 to 1 mg/L Cu |
|
Colorimetric | |
2 in 1 Test Kit |