missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Free/Total

Accurate test kits for routine water analysis

Brand:  Lovibond™ 147000

Code : 04

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10326622

  • £63.16 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Includes:
  • Aqua Comparator Housing (D55 for L147000)
  • Aqua Comparator Disc
  • DPD No. 1 Rapid Tablets
  • DPD No. 3 Rapid Tablets
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying case

Description

Description

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Specifications

±5%
Aqua comparator™ housing, aqua comparator™ disc D55, DPD No. 1 tablets, DPD No. 3 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
0.02 to 0.3mg/L
Routine water analysis
100
Chlorine Free and Total Test Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers