Learn More
Check Standards, Palintest™
Description
- Traceable reference materials defined in terms of % transmittance (%T) at a particular wavelength.
- Ideal in confirming that a Palintest™ photometer is conforming to specification.
- Each standard enables users to check each of the individual wavelengths in the photometer.
Specifications
Specifications
| Description | Check Standards Palintest |
| For Use With (Equipment) | Chlordiox-Duo |
| Type | Check Standard |