Learn More
Honeywell™ Check and Go DeepTril 1 Gloves
Product Code. 15896745
Click to view available options
Size:
7
8
9
10
Unit Size:
1 pair
Product Code. 15896745
Brand: Honeywell™ 229940010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ Check and Go DeepTril 1 Gloves are knitted in polyamide and are fully dipped in a nitrile foam coating for handling in oily or greasy conditions.
  • Black polyamide knitted glove
  • Fully dipped black foam nitrile coating with PVC dots on the palm
  • The ergonomic seamless knit construction conforms better to the natural shape of the hand for greater strength and durability
  • The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The nitrile foam and PVC dots on palm improves the grip and optimizes the glove's breathability
  • The knitted and elasticated wrist gives the glove good support
  • The nitrile coating provides outstanding abrasion resistance and impermeability to oils
  • The black color is suitable for work in a soiled atmosphere
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 Basic standards, EN 388-2003 Mechanical hazards
Description 13 gauge
Color Black
Material Foam Nitrile
Size 10
Length (Metric) 25 cm