missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CHDI 00580985

Product Code. 30448875
Change view
Click to view available options
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30448875 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30448875 Supplier Tocris Bioscience™ Supplier No. 7881/10
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective ROCK inhibitor; orally bioavailable and brain penetrant

CHDI 00580985 is a potent and selective ROCK inhibitor (IC50 values are 3 nM and 8 nM in biochemical (ROCK2) and cellular assays respectively). CHDI 00580985 inhibits ROCK-mediated substrate (MYPT1) phosphorylation (IC50 = 4 nM). In vivo, CHDI 00580985 reduces ROCK signaling in Huntington's disease mouse models in pharmacodynamic assay. CHDI 00580985 is bioavailable and brain penetrant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-4-(3-Fluoro-4-pyridinyl)-N-[(1R)-1-(3-methoxyphenyl)ethyl]-2-methyl-1-piperazinecarboxamide
CAS 2573071-18-6
Quantity 10 mg
Target Rho-kinase Inhibitors
Molecular Formula C20H25FN4O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.