missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
CHD9 Polyclonal antibody specifically detects CHD9 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | CHD9 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | AD013, ATP-dependent helicase CHD9, BC022889, CHD-9, chromatin remodeling factor CHROM1, Chromatin-related mesenchymal modulator, Chromatin-remodeling factor CHROM1, chromodomain helicase DNA binding protein 9, chromodomain-helicase-DNA-binding protein 9, ciprofibrate bound protein p240, CReMM, EC 3.6.1, EC 3.6.4.12, FLJ12178, KIAA0308, KISH2, Kismet homolog 2, Peroxisomal proliferator-activated receptor A-interacting complex 320 kDaprotein, PPAR{gamma}-interacting cofactor 320 kDa, PPAR-alpha-interacting complex protein 320 kDa, PRIC320, proteinx0008 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: SLRSQQNRNNLNPGQNSLSQSKNFMNVSGPHRVNVNHPPQMTNASNSQQSISMQQFSQTSNPSAHFHKCSSHQEGNFNGPSPNMTSCSVSNSQQFSSHYSFSSNHISPNSLLQSSA |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?