missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHD9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576529
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
CHD9 (chromodomain-helicase-DNA-binding protein 9), also known as chromatin-related mesenchymal modulator (CReMM), PPAR-α-interacting complex protein, kismet homolog 2 or CHROM1, is a 2,897 amino acid protein belonging to the Snf2/Rad54 helicase family. The CHD family of proteins are ATP-dependent chromatin remodeling enzymes which combine chromodomains with SWI2/Snf2 ATPase/helicase motifs and DNA-binding capability. Localized to the cytoplasm and the nucleus, CHD9 contains two chromodomains, one ATPbinding helicase domain and one C-terminal helicase domain. Chromodomains are protein regions of about 40-50 amino acid residues found in proteins associated with chromatin remodeling and manipulation. The domain is highly conserved among both plants and animals and is found in a large variety of proteins from many genomes. CHD9 acts as a transcriptional coactivator for PPARα and may also be an ATP-dependent chromatin remodeling protein. CHD9 is widely expressed at low levels and is present as three isoforms produced by alternative splicing.
Specifications
| CHD9 | |
| Polyclonal | |
| Unconjugated | |
| Chd9 | |
| 1810014J18Rik; 9030205D12Rik; A330063D19Rik; AD013; AD-013; ATP-dependent helicase CHD9; BC022889; Chd9; CHD-9; chromatin remodeling factor CHROM1; chromatin-related mesenchymal modulator; Chromatin-remodeling factor CHROM1; chromodomain helicase DNA binding protein 9; chromodomain-helicase-DNA-binding protein 9; ciprofibrate bound protein p240; ciprofibrate-bound protein PRIC320; CReMM; FLJ12178; Kiaa0308; KISH2; Kismet homolog 2; mKIAA0308; peroxisomal proliferator-activated receptor A-interacting complex 320 kDa protein; PPAR{gamma}-interacting cofactor 320 kDa; PPAR-alpha-interacting complex protein 320 kDa; PRIC320; proteinx0008; x0008 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109151, 80205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3L8U1, Q8BYH8 | |
| Chd9 | |
| Recombinant protein of human CHD9. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction