missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48933-25ul
This item is not returnable.
View return policy
Description
CHD9 Polyclonal antibody specifically detects CHD9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CHD9 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| AD013, ATP-dependent helicase CHD9, BC022889, CHD-9, chromatin remodeling factor CHROM1, Chromatin-related mesenchymal modulator, Chromatin-remodeling factor CHROM1, chromodomain helicase DNA binding protein 9, chromodomain-helicase-DNA-binding protein 9, ciprofibrate bound protein p240, CReMM, EC 3.6.1, EC 3.6.4.12, FLJ12178, KIAA0308, KISH2, Kismet homolog 2, Peroxisomal proliferator-activated receptor A-interacting complex 320 kDaprotein, PPAR{gamma}-interacting cofactor 320 kDa, PPAR-alpha-interacting complex protein 320 kDa, PRIC320, proteinx0008 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RNYSQSKMAHSRTSTPLLQQYQVALSASPLTSLPRLLDAKGIILEEMKVKSENLKEEPQSSEEESMSSVETRTLIKSEPVSPKNGVLPQATGDQKSGGKCETDRRMVAARTEPLTPNPA | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 80205 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction