missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD9 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60419IR
This item is not returnable.
View return policy
Description
CHD9 Polyclonal antibody specifically detects CHD9 in Human samples. It is validated for Western Blot, Immunoblot
Specifications
| CHD9 | |
| Polyclonal | |
| Western Blot, Immunoblotting | |
| AD013, ATP-dependent helicase CHD9, BC022889, CHD-9, chromatin remodeling factor CHROM1, Chromatin-related mesenchymal modulator, Chromatin-remodeling factor CHROM1, chromodomain helicase DNA binding protein 9, chromodomain-helicase-DNA-binding protein 9, ciprofibrate bound protein p240, CReMM, EC 3.6.1, EC 3.6.4.12, FLJ12178, KIAA0308, KISH2, Kismet homolog 2, Peroxisomal proliferator-activated receptor A-interacting complex 320 kDaprotein, PPAR{gamma}-interacting cofactor 320 kDa, PPAR-alpha-interacting complex protein 320 kDa, PRIC320, proteinx0008 | |
| The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human chromodomain helicase DNA binding protein 9 (chromatin-related mesenchymal modulator) using the numbering given in entry AAQ24287.1 (GeneID 80205). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoblot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 80205 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction