missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CHD8 Recombinant Protein Antigen

Product Code. 18376816 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376816 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376816 Supplier Novus Biologicals™ Supplier No. NBP317355PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 57680
Molecular Weight (g/mol) M.W. Theoretical: 27 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias ATP-dependent helicase CHD8, CHD-8, chromodomain helicase DNA binding protein 8, DKFZp686N17164, duplin, EC 3.6.1, EC 3.6.1.7, EC 3.6.4.12, Helicase with SNF2 domain 1chromodomain-helicase-DNA-binding protein 8, HELSNF1, KIAA1564axis duplication inhibitor
Gene Symbol CHD8
Research Category Wnt Signaling Pathway
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein CHD8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.