Learn More
CHD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-60418
This item is not returnable.
View return policy
Description
CHD8 Polyclonal specifically detects CHD8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| CHD8 | |
| Polyclonal | |
| 0.1 mL | |
| Store at 4C. Do not freeze. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Rabbit | |
| RUO |