missing translation for 'onlineSavingsMsg'
Learn More

CHD7 Antibody, PerCP, Novus Biologicals™

Product Code. 18409780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409780 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409780 Supplier Novus Biologicals Supplier No. NBP177393PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHD7 Polyclonal antibody specifically detects CHD7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD7
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ATP-dependent helicase CHD7, CHD-7, chromodomain helicase DNA binding protein 7, chromodomain helicase DNA binding protein 7 isoform CRA_e, chromodomain-helicase-DNA-binding protein 7, EC 3.6.1, EC 3.6.4.12, FLJ20357, FLJ20361, IS3, KIAA1416KAL5
Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human Chd7 protein (within residues 25-200). [Swiss-Prot Q9P2D1]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55636
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.