missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41133-0.025mg
This item is not returnable.
View return policy
Description
CHD7 Polyclonal specifically detects CHD7 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| CHD7 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CHD7 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human CHD7. The immunogen is located within amino acids 670 - 720 of CHD7. Amino Acid Squence: TATPKPKSSKKSSNKKPD | |
| 0.025 mg | |
| Transcription Factors and Regulators | |
| 55636 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| ATP-dependent helicase CHD7, CHD-7, chromodomain helicase DNA binding protein 7, chromodomain helicase DNA binding protein 7 isoform CRA_e, chromodomain-helicase-DNA-binding protein 7, EC 3.6.1, EC 3.6.4.12, FLJ20357, FLJ20361, IS3, KIAA1416KAL5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Multiple isoforms of CHD7 are known to exist. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction