missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
Antibody detects endogenous levels of total CHD6.
This gene encodes a member of the SNF2/RAD54 helicase protein family. The encoded protein contains two chromodomains, a helicase domain, and an ATPase domain. Several multi-subunit protein complexes remodel chromatin to allow patterns of cell type-specific gene expression, and the encoded protein is thought to be a core member of one or more of these chromatin remodeling complexes. The encoded protein may function as a transcriptional repressor and is involved in the cellular repression of influenza virus replication.
Tekniske data
Tekniske data
| Antigen | CHD6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CHD6 |
| Gene Accession No. | A3KFM7, Q8TD26 |
| Gene Alias | 5430439G14Rik; 6330406J24Rik; ATP-dependent helicase CHD6; CHD5; Chd6; CHD-6; chromodomain helicase DNA binding protein 6; chromodomain-helicase-DNA-binding protein 6; helicase C-terminal domain- and SNF2 N-terminal domain-containing protein; Kiaa1335; Radiation-induced gene B protein; RIGB |
| Gene Symbols | CHD6 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?
