missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD6 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60414JF646
This item is not returnable.
View return policy
Description
CHD6 Polyclonal antibody specifically detects CHD6 in Human samples. It is validated for Western Blot
Specifications
| CHD6 | |
| Polyclonal | |
| Western Blot | |
| ATP-dependent helicase CHD6, CHD5dJ620E11.1, CHD-6, chromodomain helicase DNA binding protein 6, EC 3.6.1, EC 3.6.4.12, FLJ22369, KIAA1335chromodomain-helicase-DNA-binding protein 6, Radiation-induced gene B protein, RIGBhelicase C-terminal domain- and SNF2 N-terminal domain-containing protein | |
| The immunogen recognized by this antibody maps to a region between residue 2665 and 2715 of human chromodomain helicase DNA binding protein 6 using the numbering given in entry NP_115597.3 (GeneID 84181). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84181 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction