missing translation for 'onlineSavingsMsg'
Learn More

CHD6 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18702901 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18702901 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702901 Supplier Novus Biologicals Supplier No. NB10060414AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHD6 Polyclonal antibody specifically detects CHD6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD6
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent helicase CHD6, CHD5dJ620E11.1, CHD-6, chromodomain helicase DNA binding protein 6, EC 3.6.1, EC 3.6.4.12, FLJ22369, KIAA1335chromodomain-helicase-DNA-binding protein 6, Radiation-induced gene B protein, RIGBhelicase C-terminal domain- and SNF2 N-terminal domain-containing protein
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2665 and 2715 of human chromodomain helicase DNA binding protein 6 using the numbering given in entry NP_115597.3 (GeneID 84181).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84181
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.