missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14173-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene belongs to the SNF2/RAD54 helicase family. It represents the main component of the nucleosome remodeling and deacetylase complex and plays an important role in epigenetic transcriptional repression. Patients with dermatomyositis develop antibodies against this protein.Specifications
| CHD4 | |
| Polyclonal | |
| Unconjugated | |
| CHD4 | |
| ATP dependent helicase CHD4, CHD 4, CHD4, Mi 2b, Mi2 BETA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 107932, 1108, 117535 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14839, Q6PDQ2 | |
| Chd4 | |
| CHD4 Fusion Protein Ag5372 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction