missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CHD4 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Specifications
Specifications
| Cell Line | HEK293 |
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Description | CHD4 HEK293 Cell Transient Overexpression Lysate(Non-Denatured) |
| Gene Accession No. | NM_001273 ;NM_001273 |
| Lysis Buffer | Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Molecular Weight (g/mol) | Theoretical M.W. 220kDa |
| Plasmid | pCMV-CHD4 full length ;pCMV-CHD4 full length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-CHD4 antibody (Mfr. No. H00001108-M01) by Western Blot. |
| Quantity | 100 μg |
| Storage Buffer | In modified RIPA Lysis Buffer. ;In modified RIPA Lysis Buffer. |
| Show More |
Safety and Handling
Product Identifier
- 100UG CHD4 HEK293 Cell Transient Overexpression Ly
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Mercaptoethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction