missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99099-100ul
This item is not returnable.
View return policy
Description
CHD4 Polyclonal antibody specifically detects CHD4 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CHD4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CHD4. | |
| 100 μg | |
| Immune System Diseases, Immunology | |
| 1108 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction