missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD4 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-57519JF646
This item is not returnable.
View return policy
Description
CHD4 Polyclonal antibody specifically detects CHD4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CHD4 | |
| Polyclonal | |
| Western Blot | |
| ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
| The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human chromodomain helicase DNA binding protein 4 (Mi2 autoantigen 218 kDa protein) using the numbering given in entry NP_001264.2 (GeneID 1108). | |
| 0.1 mL | |
| Immune System Diseases, Immunology | |
| 1108 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction