missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHD3 Recombinant Rabbit Monoclonal Antibody (23GB1380), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CHD3 |
|---|---|
| Clone | 23GB1380 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089409
![]() |
Invitrogen™
MA554070 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089372
|
Invitrogen™
MA554071 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHD3 Recombinant Monoclonal antibody specifically detects CHD3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CHD3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q12873 | |
| 1107, 216848, 303241 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1380 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2600010P09Rik; AF020312; ATP-dependent helicase CHD3; CG9594; CG9594-PA; CHD3; CHD-3; Chd3-PA; Chd7; chromodomain helicase DNA binding protein 3; chromodomain-helicase-DNA-binding protein 3; dCHD3; DmCHD3; Dmel\CG9594; Dmel_CG9594; hZFH; MGC40857; Mi 2a; Mi2 ALPHA; Mi-2 alpha; mi-2 autoantigen 240 kDa protein; Mi-2a; Mi2-alpha; Prp7; Prp9 1; Prp9-1; ZFH; Zinc finger helicase; zinc-finger helicase (Snf2-like) | |
| A synthesized peptide derived from human CHD3 (1940-2000AA). | |
| Primary | |
| -20°C | |
| CHD3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title