missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16587211
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16587211 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16587211 Supplier Invitrogen™ Supplier No. PA589256

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: A431, Jurkat, HeLa, LO2, K562; Cellular Location: Nucleus Immunogen sequence: DEFRPQNYHQ QDFRRMSDHR PAMGYHGQGP SDHYRSFHTD KLGEYKQPLP PLHPAVSDPR SPPSQKSPHD SKSPLDHRSP LERSLEQKNN PDYNWNVRKT.

The CHD family of proteins is characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHD2
Gene Accession No. O14647
Gene Alias 2810013C04Rik; 2810040A01Rik; 5630401D06Rik; AI851092; ATP-dependent helicase CHD2; BC029703; chd2; CHD-2; chromodomain helicase DNA binding protein 2; chromodomain-helicase-DNA-binding protein 2; DKFZp547I1315; DKFZp686E01200; DKFZp781D1727; Down syndrome cell adhesion molecule; EEOC; FLJ38614; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 2; si:ch211-196l7.5
Gene Symbols CHD2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1729-1828 of human CHD2 (NP_0012623).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1106
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.