missing translation for 'onlineSavingsMsg'
Learn More

CHD1L Antibody [FITC], Novus Biologicals™

Product Code. 30736550 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736550 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736550 Supplier Novus Biologicals Supplier No. AF6959F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CHD1L Polyclonal antibody specifically detects CHD1L in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD1L
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias ALC1, amplified in liver cancer 1, amplified in liver cancer protein 1, CHDL, chromodomain helicase DNA binding protein 1-like, chromodomain-helicase-DNA-binding protein 1-like, FLJ22530
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CHD1L, Arg759-Lys879, Accession # Q86WJ1
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9557
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.