missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98902CL1
This item is not returnable.
View return policy
Description
CHD1 Polyclonal antibody specifically detects CHD1 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CHD1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| ATP-dependent helicase CHD1, CHD-1, chromodomain helicase DNA binding protein 1, chromodomain-helicase-DNA-binding protein 1, DKFZp686E2337, EC 3.6.1, EC 3.6.4.12 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CHD1. | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 1105 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction