missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHCHD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534487
This item is not returnable.
View return policy
Description
A suggested positive control is rat liver tissue lysate. PA5-34487 can be used with blocking peptide PEP-1529.
CHCHD7 is a newly identified member of a multifamily of proteins containing a conserved (coiled coil 1)-(helix 1)-(coiled coil 2)-(helix 2) domain. Northern blot analysis revealed that the gene is ubiquitously expressed. Its biological function is unknown and the gene has hitherto not been associated with neoplasia. CHCHD7 and PLAG1 are located head-to-head about 500 bp apart in 8q12. A chromosomal aberration involving CHCHD7 is found in salivary gland pleiomorphic adenomas, the most common benign epithelial tumors of the salivary gland.
Specifications
| CHCHD7 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD7 | |
| 1810049H20Rik; CHCHD7; coiled-coil-helix-coiled-coil-helix domain containing 7; coiled-coil-helix-coiled-coil-helix domain-containing protein 7; COX23; COX23 cytochrome c oxidase assembly homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66433, 684258, 79145 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8K2Q5, Q9BUK0 | |
| CHCHD7 | |
| A 15 amino acid peptide near the carboxy terminus of human CHCHD7. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction