missing translation for 'onlineSavingsMsg'
Learn More

CHCHD6 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18411376 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411376 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411376 Supplier Novus Biologicals Supplier No. NBP281977IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHCHD6 Polyclonal antibody specifically detects CHCHD6 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD6
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias coiled-coil-helix-coiled-coil-helix domain containing 6, coiled-coil-helix-coiled-coil-helix domain-containing protein 6, MGC13016
Host Species Rabbit
Immunogen Rabbit polyclonal CHCHD6 antibody was raised against a 17 amino acid peptide near the amino terminus of human CHCHD6. The immunogen is located within amino acids 20 - 70 of CHCHD6. Amino Acid Squence: VNRMKEPSSPPAPTSST
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 84303
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.