missing translation for 'onlineSavingsMsg'
Learn More

CHCHD6 Antibody - BSA Free, Novus Biologicals™

Product Code. 18380731 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380731 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380731 Supplier Novus Biologicals Supplier No. NBP281977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHCHD6 Polyclonal specifically detects CHCHD6 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD6
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, ELISA
Formulation PBS
Gene Alias coiled-coil-helix-coiled-coil-helix domain containing 6, coiled-coil-helix-coiled-coil-helix domain-containing protein 6, MGC13016
Gene Symbols CHCHD6
Host Species Rabbit
Immunogen Rabbit polyclonal CHCHD6 antibody was raised against a 17 amino acid peptide near the amino terminus of human CHCHD6. The immunogen is located within amino acids 20 - 70 of CHCHD6. Amino Acid Squence: VNRMKEPSSPPAPTSST
Molecular Weight of Antigen 26 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84303
Test Specificity CHCHD6 is predicted to not cross-react to other members of the CHCHD family.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.