missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHCHD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558316
This item is not returnable.
View return policy
Description
Immunogen sequence: MQAALEVTAR YCGRELEQYG QCVAAKPESW QRDCHYLKMS IAQCTSSHPI IRQIRQACAQ PFEAFEECLR QNEAAVGNCA EHMRRFLQCA EQVQPPRSPA T Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
CHCHD5 (Coiled-Coil-Helix-Coiled-Coil-Helix Domain Containing 5) is a Protein Coding gene. Among its related pathways are Metabolism of proteins and Mitochondrial protein import.
Specifications
| CHCHD5 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD5 | |
| 1110027L01Rik; AW045710; C2orf9; Chchd5; coiled-coil-helix-coiled-coil-helix domain containing 5; coiled-coil-helix-coiled-coil-helix domain-containing protein 5; MIC14; mitochondrial intermembrane space cysteine motif protein of 14 kDa homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84269 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BSY4 | |
| CHCHD5 | |
| Recombinant protein corresponding to Human CHCHD5. Recombinant protein control fragment (Product #RP-96070). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction