missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21090-1-AP-20UL
This item is not returnable.
View return policy
Description
CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX(9)C-CX(9)C- motif in the C terminus.Specifications
| CHCHD4 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD4 | |
| CHCHD4, MIA40 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 131474, 312559, 72170 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N4Q1, Q8VEA4 | |
| Chchd4 | |
| CHCHD4 Fusion Protein Ag15339 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction