Learn More
CHCHD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25625-1-AP-150UL
 View more versions of this product
Product Code. 16834195
  • £349.00 / 150µL
Description
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintece of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintece of the MICOS complex stability and the mitochondrial cristae morphology.
Specifications
Specifications
CHCHD3
Polyclonal
Unconjugated
CHCHD3
CHCHD3, MIC19, MINOS3
Rabbit
Antigen Affinity Chromatography
RUO
54927, 66075
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CRB9, Q9NX63
CHCHD3
CHCHD3 Fusion Protein Ag22579
150 μL
Primary
Human, Mouse