missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHCHD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531578
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji, mouse liver. Predicted reactivity: Mouse (89%), Rat (83%), Pig (91%), Sheep (89%), Rhesus Monkey (95%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintenance of the MICOS complex stability and the mitochondrial cristae morphology.
Specifications
| CHCHD3 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD3 | |
| 0610041L09Rik; 1700039J09Rik; AW558177; CHCHD3; coiled-coil-helix-coiled-coil-helix domain containing 3; Coiled-coil-helix-coiled-coil-helix domain-containing protein 3; coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; Mic19; MICOS complex subunit MIC19; MINOS3; mitochondrial inner membrane organizing system 3; PPP1R22; protein phosphatase 1, regulatory subunit 22 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54927, 66075 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9CRB9, Q9NX63 | |
| CHCHD3 | |
| Recombinant protein encompassing a sequence within the center region of human CHCHD3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction