missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHCHD3 Monoclonal Antibody (24GB6560)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA557362

 View more versions of this product

Product Code. 30274885

  • £172.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CHCHD3 Monoclonal Antibody for Western Blot

Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintenance of the MICOS complex stability and the mitochondrial cristae morphology.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHCHD3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9CRB9, Q9NX63
CHCHD3
Recombinant protein of human CHCHD3 (aa 2-227).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Western Blot, Immunocytochemistry
24GB6560
Unconjugated
CHCHD3
0610041L09Rik; 1700039J09Rik; AW558177; CHCHD3; coiled-coil-helix-coiled-coil-helix domain containing 3; Coiled-coil-helix-coiled-coil-helix domain-containing protein 3; coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; Mic19; MICOS complex subunit MIC19; MINOS3; mitochondrial inner membrane organizing system 3; PPP1R22; protein phosphatase 1, regulatory subunit 22
Mouse
Affinity chromatography
RUO
296966, 54927, 66075
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.