missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHCHD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523564
This item is not returnable.
View return policy
Description
CHCHD2 Polyclonal Antibody for Western Blot, IHC (P), Flow
Transcription factor. Binds to the oxygen responsive element of COX4I2 and activates its transcription under hypoxia conditions (4% oxygen), as well as normoxia conditions (20% oxygen).
Specifications
| CHCHD2 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD2 | |
| 16.7kD protein; AAG10; Aging-associated gene 10 protein; AL033347; C7orf17; CHCHD2; coiled-coil-helix-coiled-coil-helix domain containing 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2, mitochondrial; ethanol induced 6; Etohi6; HCV NS2 trans-regulated protein; mitochondria nuclear retrograde regulator 1; MNRR1; NS2TP; PARK22 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51142 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9Y6H1 | |
| CHCHD2 | |
| KLH conjugated synthetic peptide between 74-103 amino acids from the central region of human CHCHD2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction