missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11728-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| CHCHD1 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD1 | |
| C10orf34, C2360, CHCHD1, Nuclear protein C2360 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 118487, 361005, 66121 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96BP2, Q9CQA6 | |
| Chchd1 | |
| CHCHD1 Fusion Protein Ag2325 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction