missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ CHCHD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558635
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Immunogen sequence: MATPSLRGRL ARFGNPRKPV LKPNKPLILA NRVGERRREK GEATCITEMS VMMACWKQN Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
CHCHD1 (Coiled-Coil-Helix-Coiled-Coil-Helix Domain Containing 1) is a Protein Coding gene. Among its related pathways are Mitochondrial translation and Organelle biogenesis and maintenance.
Tekniske data
| CHCHD1 | |
| Polyclonal | |
| Unconjugated | |
| Chchd1 | |
| 1110001O19Rik; 2400010G13Rik; 28S ribosomal protein S37, mitochondrial; C10orf34; C2360; CHCHD1; coiled-coil-helix-coiled-coil-helix domain containing 1; coiled-coil-helix-coiled-coil-helix domain-containing protein 1; Mitochondrial small ribosomal subunit protein mS37; MRPS37; MRP-S37; Nuclear protein C2360; similar to coiled-coil-helix-coiled-coil-helix domain containing 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 118487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96BP2 | |
| Chchd1 | |
| Recombinant protein corresponding to Human CHCHD1. Recombinant protein control fragment (Product #RP-97165). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion