missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChAT Recombinant Rabbit Monoclonal Antibody (HL2014), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA547259
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
ChAT catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses.Specifications
ChAT | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P28329, Q03059 | |
CHAT | |
Recombinant protein encompassing a sequence within the N-terminus region of human Choline Acetyltransferase. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
HL2014 | |
Unconjugated | |
CHAT | |
acetyl CoA; acetyl CoA:choline O-acetyltransferase; B230380D24Rik; CHAT; CHOACTASE; choline acetylase; choline acetyltransferase; choline acetyltransferase (EC 2.3.1.6); choline acetyltransferase long variant; choline acetyltransferase short variant; choline O-acetyltransferase; CMS1A; CMS1A2; CMS6; LOW QUALITY PROTEIN: choline O-acetyltransferase; N-type choline acetyltransferase | |
Rabbit | |
Protein A | |
RUO | |
1103, 12647 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |