Learn More
Sanyo™ Chart Paper
Product Code. 12978714
Change view
Click to view available options
Time Range:
1/7/30 Days
7 days
For Use With (Equipment):
For Use With: Chest Freezer RFT-530-010F
MTR-G85 Chart Recorder
RP-G04, MTR-G04 Chart Recorder
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Time Range For Use With (Equipment) Unit Size
12978714 1/7/30 Days RP-G04, MTR-G04 Chart Recorder Each
12928744 7 days For Use With: Chest Freezer RFT-530-010F -
12938744 7 days MTR-G85 Chart Recorder -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Please note: These goods are for sale to the UK market only.
Product Code. 12978714 Supplier Sanyo™ Supplier No. RPG04PW
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Please note: These goods are for sale to the UK market only.
Circular chart recorder paper disc. Sanyo™ Chart Paper is designed to record refrigerator/freezer temperature.
  • Circular/disc format temperature recording chart
  • Suitable for record keeping
Specifications
Product Type Chart Paper
For Use With (Equipment) RP-G04, MTR-G04 Chart Recorder
Shape Circular
Time Range 1/7/30 Days