Learn More
Sanyo™ Chart Paper
Product Code. 12928744
Click to view available options
Time Range:
1/7/30 Days
7 days
For Use With (Equipment):
For Use With: Chest Freezer RFT-530-010F
MTR-G85 Chart Recorder
RP-G04, MTR-G04 Chart Recorder
Unit Size:
Each
Please note: These goods are for sale to the UK market only.
Product Code. 12928744
Brand: Sanyo™ RP40 PW
Only null left
Add to basket
Add to basket
Please note: These goods are for sale to the UK market only.
Circular chart recorder paper disc. Sanyo™ Chart Paper is designed to record refrigerator/freezer temperature.
  • Circular/disc format temperature recording chart
  • Suitable for record keeping
Specifications
Product Type Chart Paper
For Use With (Equipment) For Use With: Chest Freezer RFT-530-010F
Shape Circular
Time Range 7 days