missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chargeswitch™ Pro ET Removal Wash Buffer

Product Code. 10308624
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10308624 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10308624 Supplier Invitrogen™ Supplier No. CS31110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Chargeswitch™ Pro ET Removal Wash Buffer is a component in the Chargeswitch™ Pro Filter Plasmid kits.

Chargeswitch™ Pro ET Removal Wash Buffer is a component in the Chargeswitch™ Pro Filter Plasmid kits. With the endotoxin removal buffer, the endotoxin levels of <10 EU/ug can be achievable and it can be used in all Chargeswitch™ Pro plasmids.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Wash Buffer
Content And Storage 500 mL Wash Buffer. Store (at room temperature)
Isolation Technology ChargeSwitch Chemistry
Sample Type DNA (Plasmid)
Final Product Type Plasmid DNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line ChargeSwitch
Product Type Pro ET Removal Wash Buffer
Quantity 1 each
Shipping Condition Room Temperature
System Type ChargeSwitch™
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.