missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Charcoal, Technical, Decolourising Powder, ActIVated, Fisher Chemical™
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Melting Point | 3652°C |
| Color | Black |
| Packaging | Plastic keg |
| Quantity | 5 kg |
| Formula Weight | 12g/mol |
| Physical Form | Powder |
Safety and Handling
Product Identifier
- Charcoal, Activated
- Warning
- Self-heating substance Category 2
- H252-Self-heating in large quantities; may catch fire.
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?