missing translation for 'onlineSavingsMsg'
Learn More

CHAPSO , MP Biomedicals

Product Code. 17801391 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17801391 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17801391 Supplier MP Biomedicals Supplier No. 0219032005
  • ONLINE EXCLUSIVE PRICE
    £646.00
    £605.85 / 5g
    Save £40.15
    6% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75621-03-3
Quantity 5 g
Molecular Formula C32H58N2O7S
Synonym 3-[(3-Cholamidopropyl)-dimethyl- ammonio]-2-hydroxy-1-propane sulfonate
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
Molecular Weight (g/mol) 614.883
PubChem CID 134129639
Formula Weight 630.9
Chemical Name or Material CHAPSO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.