missing translation for 'onlineSavingsMsg'
Learn More

Chapso, ≥98%, MP Biomedicals™

Product Code. 11491851 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
500 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491851 1 g 1g
11411861 500 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491851 Supplier MP Biomedicals Supplier No. 0219032001
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 82473-24-3
Molecular Formula C32H58N2O8S
Molecular Weight (g/mol) 630.882
InChI Key GUQQBLRVXOUDTN-AKMBKZMMSA-N
Synonym 3-[(3-Cholamidopropyl)-dimethyl- ammonio]-2-hydroxy-1-propane sulfonate
PubChem CID 134129643
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8S,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]-2-hydroxypropane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CC(CS(=O)(=O)[O-])O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range ≥98%
Formula Weight 630.9
Chemical Name or Material Chapso
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.