missing translation for 'onlineSavingsMsg'
Learn More

CHAPS, 98+%

Product Code. 11405017
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405017 1 g 1g
11415017 5 g 5g
11425017 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405017 Supplier Thermo Scientific Alfa Aesar Supplier No. B21927.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHAPS, >-98%, C32H58N2O7S, CAS Number-75621-03-3, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 25g, 614.88, H315-H319-H335, GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. | CAS: 75621-03-3 | C32H58N2O7S | 614.883 g/mol

Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.

Solubility
Slightly soluble in water.

Notes
Store in a cool. Incompatible with oxidizing agent, strong acid and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75621-03-3
Molecular Formula C32H58N2O7S
Molecular Weight (g/mol) 614.883
MDL Number MFCD00012116
InChI Key UMCMPZBLKLEWAF-GBSSQDIOSA-N
Synonym 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
PubChem CID 134129639
IUPAC Name 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
SMILES CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Specifications

Melting Point 156°C to 158°C (decomposition)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 614.88
Percent Purity ≥98%
Chemical Name or Material CHAPS
compliance-icons
Product Identifier
  • CHAPS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: CHAPS

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.