missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford CHAPS
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
1 kg
Unit Size:
10g
1g
1kg
25g
5g
Description
Zwitterionic, non-denaturing detergent for solubilizing membrane proteins.
- CAS Number: 75621-03-3
- Molecular Formula: C32H58N2SO7
- Molecular Weight: 614.9
Specifications
Specifications
| CAS | 75621-03-3 |
| Molecular Formula | C32H58N2O7S |
| Quantity | 10 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction